Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Speg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001085370.1 | NP_001078839.1 | 3 | 1472 | ||
NM_001085371.1 | NP_001078840.1 | 6 | 1939 | |||
NM_007463.4 | NP_031489.4 | 5 | 2372 | |||
GenBank mRNA | AF215896.1 | - | 5 | 2372 | ||
AK147475.1 | - | 3 | 1477 | |||
AK164360.1 | - | 6 | 1918 | |||
BC062643.1 | - | 1 | 64 |