Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aqp7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007473.4 | NP_031499.1 | 4 | 600 | ||
GenBank mRNA | AB010100.1 | - | 4 | 491 | ||
AB056091.1 | - | 4 | 434 | |||
AK076642.1 | - | 4 | 568 | |||
AK131744.1 | - | 4 | 568 | |||
BC022223.1 | - | 2 | 230 | 4242417 |