Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK018235.1 | - | 1 | 278 | ||
AK031259.1 | - | 5 | 1041 | |||
AK042977.1 | - | 5 | 1081 | |||
AK053123.1 | - | 5 | 1499 | |||
AK077591.1 | - | 5 | 1054 | |||
AK167133.1 | - | 5 | 1051 | |||
AK167601.1 | - | 5 | 1055 | |||
AK167653.1 | - | 5 | 1054 | |||
AK169094.1 | - | 5 | 1051 | |||
AK169412.1 | - | 5 | 1051 | |||
AK208914.1 | - | 1 | 75 | |||
BC010487.1 | - | 5 | 1334 | 3483748 | ||
D87899.1 | - | 5 | 1046 |