Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Astn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001205204.1 | NP_001192133.1 | 1 | 547 | ||
NM_007495.4 | NP_031521.2 | 1 | 547 | |||
GenBank mRNA | AB093222.2 | - | 1 | 133 | ||
AK045561.1 | - | 1 | 302 | |||
AK138576.1 | - | 1 | 315 | |||
AK147450.1 | - | 1 | 317 | |||
AK164117.1 | - | 1 | 512 | |||
AK165200.1 | - | 1 | 300 | |||
BC094666.1 | - | 1 | 433 | 30545499 | ||
CF746126.1 | - | 1 | 547 |