Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Astn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001205204.1 | NP_001192133.1 | 3 | 1051 | ||
NM_007495.4 | NP_031521.2 | 3 | 1051 | |||
GenBank mRNA | AB093222.2 | - | 3 | 637 | ||
AK045561.1 | - | 3 | 806 | |||
AK138576.1 | - | 3 | 819 | |||
AK147450.1 | - | 3 | 821 | |||
AK164117.1 | - | 3 | 1016 | |||
AK165200.1 | - | 3 | 804 | |||
BC094666.1 | - | 3 | 937 | 30545499 | ||
G49230.1 | - | 3 | 639 | |||
U48797.1 | - | 3 | 639 |