Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atp1b3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007502.4 | NP_031528.1 | 5 | 705 | ||
GenBank mRNA | AK032516.1 | - | 5 | 722 | ||
AK148495.1 | - | 3 | 410 | |||
AK151700.1 | - | 5 | 722 | |||
AK151841.1 | - | 5 | 721 | |||
AK159532.1 | - | 5 | 722 | |||
AK161282.1 | - | 5 | 737 | |||
AK168273.1 | - | 5 | 721 | |||
AK177007.1 | - | 2 | 210 | |||
AK191321.1 | - | 2 | 159 | |||
AK218119.1 | - | 3 | 302 | |||
BC079916.1 | - | 5 | 710 | 30665138 | ||
BC114530.1 | - | 2 | 130 | |||
U59761.1 | - | 5 | 708 |