Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc7a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK077593.1 | - | 11 | 1796 | ||
AK141558.1 | - | 10 | 2268 | |||
AK165931.1 | - | 11 | 1793 | |||
BC050195.1 | - | 11 | 1772 | 30010078 | ||
U70859.1 | - | 12 | 1930 |