Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bcap29 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164090.1 | NP_001157562.1 | 8 | 1243 | ||
NM_007530.3 | NP_031556.1 | 7 | 1406 | |||
GenBank mRNA | AK090068.1 | - | 8 | 1220 | ||
AK144754.1 | - | 8 | 1217 | |||
AK218893.1 | - | 1 | 357 | |||
BC003303.1 | - | 7 | 1310 | 3584161 | ||
BC021661.1 | - | 7 | 1417 | 2646883 | ||
BC040751.1 | - | 2 | 1596 | |||
X78684.1 | - | 7 | 1217 |