Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Phb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007531.2 | NP_031557.2 | 7 | 884 | ||
GenBank mRNA | AK011107.1 | - | 7 | 851 | ||
AK090088.1 | - | 7 | 841 | |||
AK132052.1 | - | 7 | 846 | |||
AK132058.1 | - | 7 | 873 | |||
AK135729.1 | - | 7 | 801 | |||
AK151952.1 | - | 7 | 878 | |||
AK152375.1 | - | 6 | 1533 | |||
AK153368.1 | - | 6 | 1533 | |||
AY211613.1 | - | 7 | 846 | |||
AY319418.1 | - | 7 | 736 | |||
BC145875.1 | - | 7 | 836 | 40131178 | ||
X78683.1 | - | 7 | 840 |