Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bgn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007542.4 | NP_031568.2 | 6 | 876 | ||
GenBank mRNA | AK135496.1 | - | 6 | 877 | ||
AK155973.1 | - | 6 | 877 | |||
AK164867.1 | - | 6 | 877 | |||
AK171870.1 | - | 6 | 877 | |||
BC005452.1 | - | 6 | 870 | 3594469 | ||
BC019502.1 | - | 6 | 879 | 4237268 | ||
BC052857.1 | - | 6 | 861 | 30061850 | ||
L20276.1 | - | 6 | 876 | |||
X53928.1 | - | 971 |