Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cxcr5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007551.2 | NP_031577.2 | 2 | 685 | ||
GenBank mRNA | AK089611.1 | - | 2 | 702 | ||
AK133064.1 | - | 4 | 917 | |||
AK155954.1 | - | 2 | 682 | |||
AK156087.1 | - | 2 | 682 | |||
AK157016.1 | - | 2 | 682 | |||
BC064059.1 | - | 2 | 685 | 1366773 | ||
L20332.1 | - | 1 | 192 | |||
X71788.1 | - | 2 | 642 |