Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bmp5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007555.3 | NP_031581.2 | 2 | 1348 | ||
GenBank mRNA | AK033362.1 | - | 2 | 1351 | ||
AK160412.1 | - | 456 | ||||
BC100751.1 | - | 2 | 613 | |||
BC100752.1 | - | 2 | 613 | |||
BC100753.1 | - | 2 | 619 | |||
BC100754.1 | - | 2 | 613 | |||
BC141283.1 | - | 2 | 1145 | 9055895 | ||
L41145.1 | - | 2 | 614 |