Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bmp8b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007559.4 | NP_031585.2 | 4, 5 | 1040 | ||
GenBank mRNA | AK038590.1 | - | 262 | |||
AK082895.1 | - | 4, 5 | 1041 | |||
BC137890.1 | - | 4, 5 | 910 | 8860909 | ||
U39545.1 | - | 4, 5 | 1040 |