Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bmpr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007561.4 | NP_031587.1 | 2 | 476 | ||
GenBank mRNA | AF003942.1 | - | 2 | 476 | ||
AK149385.1 | - | 1357 | ||||
U78048.1 | - | 2 | 118 |