Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other COL13A1 SIRNA Assays ›
Gene Symbol:
COL13A1
Gene Name:
collagen type XIII alpha 1 chain
Gene Aliases:
CMS19, COLXIIIA1
Chromosome Location:
Chr.10: 69801885 - 69959148 on Build GRCh38
UniGene ID:
Hs.695934
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
15989
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: COL13A1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001130103.1
NP_001123575.1
7
1040
NM_001320951.1
NP_001307880.1
8
1067
NM_080798.3
NP_542988.3
4
926
NM_080800.3
NP_542990.3
7
1040
NM_080801.3
NP_542991.3
7
1040
NM_080802.3
NP_542992.3
7
1040
NM_080805.3
NP_542995.3
5
953
XM_011539292.2
XP_011537594.1
8
1070
XM_011539293.2
XP_011537595.1
8
1070
XM_011539294.2
XP_011537596.1
8
1070
XM_011539295.2
XP_011537597.1
8
1070
XM_017015676.1
XP_016871165.1
8
1070
XM_017015677.1
XP_016871166.1
8
1070
XM_017015678.1
XP_016871167.1
7
1043
XM_017015679.1
XP_016871168.1
8
1070
XM_017015680.1
XP_016871169.1
8
1070
XM_017015681.1
XP_016871170.1
8
1070
XM_017015682.1
XP_016871171.1
8
1070
XM_017015683.1
XP_016871172.1
7
1034
XM_017015684.1
XP_016871173.1
7
1043
XM_017015685.1
XP_016871174.1
8
1070
XM_017015686.1
XP_016871175.1
8
1070
XM_017015687.1
XP_016871176.1
8
1070
XM_017015688.1
XP_016871177.1
8
1070
XM_017015689.1
XP_016871178.1
8
1070
XM_017015690.1
XP_016871179.1
5
980
XM_017015691.1
XP_016871180.1
8
1070
XM_017015692.1
XP_016871181.1
7
1034
XM_017015693.1
XP_016871182.1
8
1070
XM_017015694.1
XP_016871183.1
4
929
XM_017015695.1
XP_016871184.1
4
929
XM_017015696.1
XP_016871185.1
5
223
XM_017015697.1
XP_016871186.1
7
501
GenBank mRNA
BC136385.1
-
4
696
9020372
Back To Top
More Information
Panther Classification:
Molecular Function -
extracellular matrix structural protein
Gene Ontology Categories:
Function(s)
Process(es)
morphogenesis of a branching structure
endochondral ossification
cell-matrix adhesion
single organismal cell-cell adhesion
cell differentiation
extracellular matrix organization
collagen catabolic process
protein binding
heparin binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-cccc65c97-xbcvc:80/10.239.164.135:80.
git-commit: 2b810224bbfb944e11adb8d8c7a69f36ca15064f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.1-offline