Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bnc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007562.2 | NP_031588.2 | 4 | 541 | ||
GenBank mRNA | AF025301.1 | - | 2 | 349 | ||
AK087567.1 | - | 4 | 535 | |||
U88064.1 | - | 4 | 488 |