Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Btg2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007570.2 | NP_031596.1 | 2 | 2451 | ||
GenBank mRNA | AK133796.1 | - | 2 | 2416 | ||
AK169269.1 | - | 2 | 2416 | |||
AK171400.1 | - | 1 | 1106 | |||
BC132259.1 | - | 2 | 2402 | 40130536 | ||
BC138639.1 | - | 2 | 2400 | 8861659 |