Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C4bp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007576.3 | NP_031602.3 | 3 | 412 | ||
GenBank mRNA | AB094485.1 | - | 393 | |||
AB094486.1 | - | 3 | 328 | |||
AK136735.1 | - | 3 | 326 | |||
AK149468.1 | - | 3 | 419 | |||
AK149477.1 | - | 3 | 329 | |||
AK162345.1 | - | 3 | 326 | |||
BC012257.1 | - | 3 | 384 | 4163203 | ||
M17122.1 | - | 3 | 426 |