Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cacna1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252059.1 | NP_001238988.1 | 18 | 2635 | ||
NM_001252060.1 | NP_001238989.1 | 19 | 2776 | |||
NM_001252061.1 | NP_001238990.1 | 19 | 2776 | |||
NM_007578.3 | NP_031604.3 | 19 | 2776 | |||
GenBank mRNA | AB066608.1 | - | 19 | 2549 | ||
AB066609.1 | - | 19 | 2549 | |||
AK147277.1 | - | 18 | 2623 | |||
AK147288.1 | - | 19 | 2776 | |||
AK147498.1 | - | 18 | 2623 | |||
AK147697.1 | - | 18 | 2623 | |||
AY714490.1 | - | 19 | 2549 | |||
U76716.1 | - | 19 | 2351 |