Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Car8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007592.3 | NP_031618.2 | 7 | 786 | ||
GenBank mRNA | AK004896.1 | - | 7 | 787 | ||
AK004923.1 | - | 7 | 787 | |||
BC010773.1 | - | 7 | 762 | 4216498 | ||
EU234006.1 | - | 6 | 612 |