Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DIAPH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001079812.2 | NP_001073280.1 | 18 | 2622 | ||
NM_001314007.1 | NP_001300936.1 | 19 | 2649 | |||
NM_005219.4 | NP_005210.3 | 19 | 2649 | |||
XM_011537572.2 | XP_011535874.1 | 18 | 2590 | |||
XM_011537573.2 | XP_011535875.1 | 19 | 2457 | |||
GenBank mRNA | AB209482.1 | - | 19 | 2590 | ||
AF051782.1 | - | 18 | 2436 | |||
AK023345.1 | - | 12 | 1831 | |||
AK301195.1 | - | 4 | 385 | |||
AK307822.1 | - | 15 | 2081 | |||
AY363395.1 | - | 19 | 2508 | |||
BC007411.2 | - | 3 | 60 | |||
BC117257.1 | - | 18 | 2496 | 40125808 | ||
BC143413.1 | - | 19 | 2526 | 9051921 | ||
BC143414.1 | - | 19 | 2526 | |||
DQ067452.1 | - | 4 | 660 | |||
DQ067453.1 | - | 18 | 2493 |