Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301067.1 | NP_001287996.1 | 7 | 812 | ||
NM_007598.4 | NP_031624.2 | 7 | 796 | |||
GenBank mRNA | AK053351.1 | - | 7 | 620 | ||
AK128889.1 | - | 7 | 618 | |||
AK137222.1 | - | 7 | 601 | |||
AK144094.1 | - | 7 | 604 | |||
AK154002.1 | - | 7 | 816 | |||
AK156556.1 | - | 7 | 630 | |||
AK158150.1 | - | 6 | 1460 | |||
AK165565.1 | - | 7 | 599 | |||
AK169652.1 | - | 7 | 591 | |||
AK170901.1 | - | 7 | 607 | |||
AK171388.1 | - | 7 | 614 | |||
BC005446.1 | - | 6 | 595 | 3600514 | ||
BC005472.1 | - | 6 | 588 | 3590890 | ||
L12367.1 | - | 7 | 622 |