Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Capn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001110504.1 | NP_001103974.1 | 4 | 473 | ||
NM_007600.3 | NP_031626.1 | 4 | 525 | |||
GenBank mRNA | AF021847.1 | - | 4 | 404 | ||
AF084459.1 | - | 3 | 391 | |||
AK088547.1 | - | 4 | 445 | |||
AK149706.1 | - | 4 | 465 | |||
AK150084.1 | - | 4 | 464 | |||
AK160940.1 | - | 4 | 465 | |||
AK165258.1 | - | 4 | 484 | |||
AK168063.1 | - | 4 | 464 | |||
AK171405.1 | - | 4 | 428 | |||
BC026138.1 | - | 4 | 440 | 4218148 | ||
BC050276.1 | - | 4 | 441 | 4923347 |