Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BRDT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242805.2 | NP_001229734.2 | 5, 6 | 855 | ||
NM_001242807.2 | NP_001229736.2 | 3, 4 | 454 | |||
NM_001242808.2 | NP_001229737.2 | 3, 4 | 648 | |||
NM_001242810.2 | NP_001229739.2 | 4, 5 | 1142 | |||
NM_001726.4 | NP_001717.3 | 4, 5 | 581 | |||
NM_207189.3 | NP_997072.2 | 4, 5 | 786 | |||
XM_006710853.3 | XP_006710916.1 | 5, 6 | 1371 | |||
XM_006710854.3 | XP_006710917.1 | 6, 7 | 1216 | |||
XM_006710855.3 | XP_006710918.1 | 6, 7 | 1221 | |||
XM_006710856.3 | XP_006710919.1 | 6, 7 | 1195 | |||
XM_006710857.3 | XP_006710920.1 | 6, 7 | 1185 | |||
XM_011542032.2 | XP_011540334.1 | 6, 7 | 705 | |||
XM_011542033.2 | XP_011540335.1 | 6, 7 | 679 | |||
XM_011542034.2 | XP_011540336.1 | 6, 7 | 695 | |||
XM_011542035.2 | XP_011540337.1 | 6, 7 | 669 | |||
XM_011542036.2 | XP_011540338.1 | 4, 5 | 584 | |||
GenBank mRNA | AF019085.1 | - | 4, 5 | 544 | ||
AK302758.1 | - | 3, 4 | 390 | |||
AK316442.1 | - | 3, 4 | 453 | |||
AY338951.1 | - | 5, 6 | 855 | |||
BC005281.1 | - | 4, 5 | 512 | |||
BC017582.1 | - | 4, 5 | 587 | |||
BC047900.1 | - | 4, 5 | 551 | |||
BC062700.1 | - | 4, 5 | 546 | |||
BG717291.1 | - | 4, 5 | 577 | |||
BX648135.1 | - | 4, 5 | 877 |