Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Casp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK042072.1 | - | 2 | 186 | ||
AK052396.1 | - | 1 | 2538 | |||
AK133829.1 | - | 2 | 195 | |||
AK197928.1 | - | 1 | 114 | |||
AK210521.1 | - | 2 | 114 | |||
BC034262.1 | - | 2 | 252 | 4460089 | ||
BY074786.1 | - | 2 | 308 | |||
D28492.1 | - | 2 | 175 | |||
Y13085.1 | - | 2 | 254 |