Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Casp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK042072.1 | - | 11 | 3383 | ||
AK133829.1 | - | 11 | 3395 | |||
AK142569.1 | - | 4 | 2525 | |||
AK170649.1 | - | 10 | 3116 | |||
AK196678.1 | - | 1 | 210 | |||
AK199486.1 | - | 1 | 210 | |||
AK200675.1 | - | 1 | 210 | |||
AK210714.1 | - | 1 | 210 | |||
AK214788.1 | - | 1 | 211 | |||
BC034262.1 | - | 11 | 3450 | 4460089 | ||
D28492.1 | - | 11 | 3373 | |||
Y13085.1 | - | 11 | 3452 |