Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cbl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007619.2 | NP_031645.2 | 2 | 399 | ||
GenBank mRNA | AK029826.1 | - | 2 | 402 | ||
AK153915.1 | - | 2 | 351 | |||
AK163907.1 | - | 2 | 350 | |||
BC125285.1 | - | 2 | 314 | 40129800 | ||
X57111.1 | - | 2 | 337 |