Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cckbr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF019371.1 | - | 2 | 172 | ||
AK028325.1 | - | 2 | 382 | |||
AK039721.1 | - | 381 | ||||
AK053307.1 | - | 2 | 242 | |||
BC103530.1 | - | 2 | 352 | 40039482 | ||
BC103531.1 | - | 2 | 352 | 40039474 | ||
BC103537.1 | - | 3 | 297 | |||
BC103538.1 | - | 2 | 352 | 40039475 |