Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccnd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007631.2 | NP_031657.1 | 5 | 1725 | ||
GenBank mRNA | AK017834.1 | - | 5 | 1687 | ||
AK053348.1 | - | 5 | 1633 | |||
AK149905.1 | - | 5 | 1632 | |||
AK164997.1 | - | 5 | 1670 | |||
AK169905.1 | - | 1 | 562 | |||
AK170833.1 | - | 5 | 1609 | |||
BC044841.1 | - | 5 | 1631 | 3155470 | ||
S78355.1 | - | 5 | 1633 |