Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccne1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007633.2 | NP_031659.2 | 8 | 833 | ||
GenBank mRNA | AK089950.1 | - | 8 | 765 | ||
AK163046.1 | - | 8 | 822 | |||
AK165658.1 | - | 8 | 823 | |||
BC003499.1 | - | 4 | 385 | |||
BC062152.1 | - | 8 | 860 | 6510163 | ||
BC084588.1 | - | 8 | 877 | 6506897 | ||
BC106191.1 | - | 8 | 833 | 6336006 | ||
BC138660.1 | - | 7 | 666 | 8861680 | ||
BC138662.1 | - | 7 | 666 | 8861683 | ||
X75888.1 | - | 7 | 673 |