Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cct5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007637.2 | NP_031663.1 | 4 | 494 | ||
GenBank mRNA | AK077753.1 | - | 4 | 452 | ||
AK088184.1 | - | 4 | 428 | |||
AK129056.1 | - | 4 | 400 | |||
AK133857.1 | - | 4 | 452 | |||
AK136493.1 | - | 4 | 446 | |||
AK145445.1 | - | 4 | 463 | |||
AK146405.1 | - | 4 | 466 | |||
AK153911.1 | - | 4 | 470 | |||
AK167895.1 | - | 4 | 435 | |||
AK217822.1 | - | 3 | 247 | |||
BC092233.1 | - | 4 | 464 | |||
BC094427.1 | - | 4 | 441 | 30537966 | ||
Z31555.1 | - | 4 | 494 |