Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ms4a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007641.5 | NP_031667.1 | 5 | 586 | ||
GenBank mRNA | AK017903.1 | - | 5 | 587 | ||
AK041428.1 | - | 5 | 585 | |||
AK080040.1 | - | 5 | 552 | |||
AK080148.1 | - | 584 | ||||
AK137469.1 | - | 5 | 575 | |||
AK143824.1 | - | 4 | 2464 | |||
AK156033.1 | - | 585 | ||||
AK166571.1 | - | 5 | 680 | |||
AK166659.1 | - | 5 | 680 | |||
AK171631.1 | - | 5 | 615 | |||
AK171638.1 | - | 5 | 538 | |||
AK171659.1 | - | 5 | 571 | |||
BC028322.1 | - | 5 | 567 | 4503841 | ||
CT010253.1 | - | 4 | 467 | |||
M62541.1 | - | 644 |