Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ms4a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007641.5 | NP_031667.1 | 7 | 1939 | ||
GenBank mRNA | AK017903.1 | - | 7 | 1940 | ||
AK041428.1 | - | 7 | 1938 | |||
AK080148.1 | - | 1937 | ||||
AK156033.1 | - | 1938 | ||||
AK166571.1 | - | 7 | 2035 | |||
AK166659.1 | - | 7 | 2035 | |||
AK171631.1 | - | 7 | 1968 | |||
AK171638.1 | - | 7 | 1892 |