Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Entpd5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001026214.2 | NP_001021385.1 | 4 | 444 | ||
NM_001286049.1 | NP_001272978.1 | 3 | 419 | |||
NM_001286058.1 | NP_001272987.1 | 2 | 346 | |||
NM_007647.3 | NP_031673.2 | 3 | 371 | |||
GenBank mRNA | AF006482.1 | - | 3 | 379 | ||
AF136571.1 | - | 3 | 405 | |||
AJ238636.1 | - | 3 | 407 | |||
AK002618.1 | - | 3 | 344 | |||
AK031581.1 | - | 4 | 424 | |||
AK036641.1 | - | 4 | 421 | |||
AK037736.1 | - | 3 | 340 | |||
AK045828.1 | - | 3 | 357 | |||
AK079267.1 | - | 2 | 268 | |||
AK080265.1 | - | 4 | 431 | |||
AK081435.1 | - | 4 | 409 | |||
AK088455.1 | - | 3 | 400 | |||
AK160950.1 | - | 1 | 265 | |||
AK163688.1 | - | 3 | 345 | |||
AW012371.1 | - | 3 | 419 | |||
BC015247.1 | - | 2 | 329 | 4242560 |