Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cd63 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042580.1 | NP_001036045.1 | 6 | 595 | ||
NM_001282966.1 | NP_001269895.1 | 6 | 639 | |||
NM_007653.3 | NP_031679.1 | 6 | 554 | |||
GenBank mRNA | AF159427.1 | - | 6 | 475 | ||
AK028220.1 | - | 6 | 492 | |||
AK146618.1 | - | 6 | 542 | |||
AK151702.1 | - | 6 | 520 | |||
AK151963.1 | - | 6 | 494 | |||
AK152072.1 | - | 6 | 516 | |||
AK152372.1 | - | 6 | 520 | |||
AK159875.1 | - | 6 | 517 | |||
AK163785.1 | - | 6 | 516 | |||
AK166373.1 | - | 1 | 2269 | |||
AK166543.1 | - | 6 | 515 | |||
AK166766.1 | - | 6 | 514 | |||
AK170344.1 | - | 6 | 549 | |||
AK182243.1 | - | 4 | 350 | |||
AK210314.1 | - | 4 | 352 | |||
AK216882.1 | - | 4 | 350 | |||
BC008108.1 | - | 6 | 501 | 3157993 | ||
BC012212.1 | - | 6 | 501 | 3157993 | ||
BC083161.1 | - | 6 | 486 | 6530532 | ||
CJ210573.1 | - | 1 | 31 | |||
CT010179.1 | - | 5 | 450 | |||
D16432.1 | - | 6 | 504 |