Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cd72 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001110320.1 | NP_001103790.1 | 6 | 912 | ||
NM_001110321.1 | NP_001103791.1 | 5 | 840 | |||
NM_001110322.1 | NP_001103792.1 | 4 | 750 | |||
NM_007654.2 | NP_031680.2 | 6 | 912 | |||
GenBank mRNA | AA203771.1 | - | 1 | 86 | ||
AK134132.1 | - | 6 | 915 | |||
AK144191.1 | - | 5 | 843 | |||
AK151094.1 | - | 752 | ||||
J04170.1 | - | 6 | 849 | |||
S40777.1 | - | 6 | 845 |