Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cd82 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136055.2 | NP_001129527.1 | 8 | 763 | ||
NM_001271430.1 | NP_001258359.1 | 7 | 819 | |||
NM_001271431.1 | NP_001258360.1 | 7 | 687 | |||
NM_001271432.1 | NP_001258361.1 | 9 | 982 | |||
NM_001271461.1 | NP_001258390.1 | 8 | 774 | |||
NM_001271462.1 | NP_001258391.1 | 8 | 786 | |||
NM_007656.5 | NP_031682.1 | 8 | 895 | |||
GenBank mRNA | AK135381.1 | - | 8 | 759 | ||
AK146912.1 | - | 8 | 759 | |||
AK152871.1 | - | 8 | 890 | |||
AK153070.1 | - | 8 | 890 | |||
AK168097.1 | - | 8 | 846 | |||
AK168515.1 | - | 8 | 865 | |||
BC132559.1 | - | 8 | 865 | 40130836 | ||
BC132565.1 | - | 8 | 865 | 40130842 | ||
D14883.1 | - | 7 | 728 |