Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cd82 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136055.2 | NP_001129527.1 | 10 | 1376 | ||
NM_001271430.1 | NP_001258359.1 | 9 | 1432 | |||
NM_001271431.1 | NP_001258360.1 | 9 | 1300 | |||
NM_001271432.1 | NP_001258361.1 | 11 | 1595 | |||
NM_001271461.1 | NP_001258390.1 | 10 | 1387 | |||
NM_001271462.1 | NP_001258391.1 | 10 | 1399 | |||
NM_007656.5 | NP_031682.1 | 10 | 1508 | |||
GenBank mRNA | AK135381.1 | - | 10 | 1372 | ||
AK146912.1 | - | 10 | 1372 | |||
AK152871.1 | - | 10 | 1504 | |||
AK153070.1 | - | 10 | 1503 | |||
AK160907.1 | - | 9 | 1355 | |||
AK168097.1 | - | 10 | 1459 | |||
AK168515.1 | - | 10 | 1478 | |||
D14883.1 | - | 9 | 1340 |