Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdc25a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007658.3 | NP_031684.3 | 2, 3 | 655 | ||
GenBank mRNA | AK077609.1 | - | 2, 3 | 272 | ||
AK077991.1 | - | 2, 3 | 658 | |||
AK142334.1 | - | 1, 2 | 930 | |||
BC046296.1 | - | 2, 3 | 274 | 6486343 | ||
U27323.1 | - | 2, 3 | 241 |