Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdc25a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007658.3 | NP_031684.3 | 13 | 1610 | ||
GenBank mRNA | AK036499.1 | - | 3 | 1062 | ||
AK077609.1 | - | 13 | 1228 | |||
AK077991.1 | - | 13 | 1612 | |||
AK142334.1 | - | 12 | 1885 | |||
BC046296.1 | - | 13 | 1229 | 6486343 | ||
U27323.1 | - | 13 | 1199 |