Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdkn2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007670.4 | NP_031696.1 | 2 | 1045 | ||
GenBank mRNA | AF059567.1 | - | 2 | 953 | ||
AK144132.1 | - | 2 | 1055 | |||
AK146668.1 | - | 2 | 1007 | |||
BC002010.1 | - | 2 | 959 | 3495097 |