Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chil1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007695.3 | NP_031721.2 | 10 | 1479 | ||
GenBank mRNA | AK051475.1 | - | 10 | 1420 | ||
AK155412.1 | - | 10 | 1420 | |||
BC003780.1 | - | 10 | 1415 | 3601796 | ||
BC004734.1 | - | 10 | 1406 | 3583267 | ||
BC005611.1 | - | 10 | 1426 | 3582304 | ||
X93035.1 | - | 10 | 1420 |