Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007697.2 | NP_031723.2 | 3 | 502 | ||
GenBank mRNA | AK040765.1 | - | 432 | |||
AK053039.1 | - | 2 | 304 | |||
AK082640.1 | - | 3 | 417 | |||
BC060216.1 | - | 3 | 469 | |||
BC131670.1 | - | 3 | 310 | 40110867 | ||
BC131671.1 | - | 3 | 306 | 40110873 | ||
X94310.1 | - | 3 | 376 |