Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chuk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001162410.1 | NP_001155882.1 | 9 | 961 | ||
NM_007700.2 | NP_031726.2 | 9 | 961 | |||
GenBank mRNA | AK018671.1 | - | 9 | 940 | ||
AK035326.1 | - | 9 | 918 | |||
AK150039.1 | - | 9 | 940 | |||
AK155328.1 | - | 9 | 941 | |||
AK155393.1 | - | 9 | 962 | |||
AK164236.1 | - | 9 | 962 | |||
BC018243.1 | - | 9 | 953 | 4511275 | ||
U12473.1 | - | 9 | 929 |