Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chuk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001162410.1 | NP_001155882.1 | 21 | 3174 | ||
NM_007700.2 | NP_031726.2 | 21 | 3191 | |||
GenBank mRNA | AK008539.1 | - | 738 | |||
AK035326.1 | - | 21 | 3148 | |||
AK155393.1 | - | 21 | 3175 | |||
AK164236.1 | - | 21 | 3175 | |||
U12473.1 | - | 21 | 3159 |