Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cmah | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001111110.2 | NP_001104580.1 | 4, 5 | 693 | ||
NM_001284519.1 | NP_001271448.1 | 5, 6 | 855 | |||
NM_001284520.1 | NP_001271449.1 | 3, 4 | 348 | |||
NM_007717.5 | NP_031743.3 | 3, 4 | 348 | |||
GenBank mRNA | AB061276.1 | - | 4, 5 | 677 | ||
AB061277.1 | - | 5, 6 | 839 | |||
AK040274.1 | - | 2, 3 | 298 | |||
AK083394.1 | - | 2, 3 | 382 | |||
AK145110.1 | - | 2, 3 | 496 | |||
AK145199.1 | - | 3, 4 | 349 | |||
AK166486.1 | - | 3, 4 | 349 | |||
BC028474.1 | - | 4, 5 | 703 | |||
BC055079.1 | - | 2, 3 | 345 | 30077348 | ||
CA577904.1 | - | 2, 3 | 333 | |||
D21826.1 | - | 2, 3 | 320 |