Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HAGH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040427.1 | NP_001035517.1 | 3 | 724 | ||
NM_001286249.1 | NP_001273178.1 | 2 | 628 | |||
NM_005326.4 | NP_005317.2 | 2 | 628 | |||
XM_011522469.2 | XP_011520771.1 | 2 | 268 | |||
XM_011522470.2 | XP_011520772.1 | 2 | 266 | |||
XM_017023195.1 | XP_016878684.1 | 2 | 193 | |||
GenBank mRNA | AK290155.1 | - | 2 | 193 | ||
AK298174.1 | - | 2 | 268 | |||
AK299173.1 | - | 2 | 259 | |||
AK299993.1 | - | 2 | 268 | |||
AK308825.1 | - | 2 | 236 | |||
BC000840.1 | - | 2 | 248 | 3457214 | ||
BC002627.2 | - | 2 | 577 | 3608098 | ||
BG703401.1 | - | 2 | 560 | |||
BU176541.1 | - | 3 | 329 | |||
DB502986.1 | - | 3 | 353 | |||
DR006636.1 | - | 2 | 613 | |||
X90999.1 | - | 1 | 113 |