Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Coch | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001198835.1 | NP_001185764.1 | 12 | 2422 | ||
NM_007728.5 | NP_031754.1 | 11 | 2621 | |||
GenBank mRNA | AF006741.1 | - | 12 | 2405 | ||
AK028690.1 | - | 11 | 2508 | |||
AK137336.1 | - | 11 | 2622 | |||
AK156430.1 | - | 12 | 2387 | |||
AK171875.1 | - | 12 | 2390 | |||
BC045137.1 | - | 12 | 2372 | 6306601 |