Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HADH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001184705.2 | NP_001171634.2 | 2 | 407 | ||
NM_005327.4 | NP_005318.3 | 2 | 407 | |||
XM_005262972.2 | XP_005263029.1 | 2 | 339 | |||
GenBank mRNA | AF001902.1 | - | 2 | 171 | ||
AF001903.1 | - | 2 | 407 | |||
AK096018.1 | - | 2 | 339 | |||
AK314444.1 | - | 2 | 268 | |||
BC000306.2 | - | 2 | 215 | 2820507 | ||
DA825380.1 | - | 2 | 337 | |||
X96752.1 | - | 2 | 275 |