Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Col4a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007734.2 | NP_031760.2 | 6, 7 | 810 | ||
GenBank mRNA | AF169387.1 | - | 6, 7 | 755 |